Learn More
Bellco Glass, Inc. TC Treated 24-Well Plate W/Lid
SDP
Catalog No. 502329753
Encompass_Preferred
24-Well Plates are available non-sterile, sterile, and treated for tissue culture. Sterile and treated plates are gamma-irradiated for sterility and are individually packed with lids.Distinct alpha-numeric well designationsWell volume: 3.5 mLLid sold separately6-well, 12-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico. Style-TC Treated and Sterile, Lid-With Lid, Material-Polystyrene (PS), Color-Natural, Package Type-Individually Packed, Carton Qty-100
Catalog No. 50-232-9753 Supplier Bellco Glass, Inc. Supplier No. 130000313
24-Well Plates are available non-sterile, sterile, and treated for tissue culture. Sterile and treated plates are gamma-irradiated for sterility and are individually packed with lids.Distinct alpha-numeric well designationsWell volume: 3.5 mLLid sold separately6-well, 12-well, 96-well and 384-well units are also available - Items are not available for shipment to Mexico. Style-TC Treated and Sterile, Lid-With Lid, Material-Polystyrene (PS), Color-Natural, Package Type-Individually Packed, Carton Qty-100