Learn More
Genesee Scientific Corporation GenClone Cell Culture Dishes, 100 mm Diameter, Sterile, 300 Dishes/Unit
SDP
Catalog No. NC1121418
Encompass
GenClone Cell and Tissue Culture Dishes are designed for optimal cell adhesion and proliferation in a range of applications. These dishes feature a flat growth surface and vented lids that enhance gas exchange, ensuring uniform cell growth. Manufactured under strict sterile conditions, they are DNase, RNase free, and non-pyrogenic, making them reliable for sensitive experiments. The integrated gripping ring provides enhanced handling, differentiating them from standard dishes.
Catalog No. NC1121418 Supplier Genesee Scientific Corporation Supplier No. 25202
GenClone Cell and Tissue Culture Dishes are designed for optimal cell adhesion and proliferation in a range of applications. These dishes feature a flat growth surface and vented lids that enhance gas exchange, ensuring uniform cell growth. Manufactured under strict sterile conditions, they are DNase, RNase free, and non-pyrogenic, making them reliable for sensitive experiments. The integrated gripping ring provides enhanced handling, differentiating them from standard dishes.