Learn More
Sigma Aldrich Fine Chemicals Biosciences Corning(R) Costar(R) TC-Treated Multiple Well Plates size 24 wells, polystyrene plate, flat bottom wells, case of 100 (individually wrapped), sterile, lid | 100EA
Catalog No. NC1937461
Encompass
Corning(R) Costar(R) TC-Treated Multiple Well Plates size 24 wells, polystyrene plate, flat bottom wells, case of 100 (individually wrapped), sterile, lid | 100EA
Catalog No. NC1937461 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS3524100EA
Corning(R) Costar(R) TC-Treated Multiple Well Plates size 24 wells, polystyrene plate, flat bottom wells, case of 100 (individually wrapped), sterile, lid | 100EA