Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One TC Treated Terasaki Plate
GREENER_CHOICE

Catalog No. 07000624 Shop All Greiner Bio-One Products
Change view
Click to view available options
Well Count:
60
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Count
07-000-624 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-624 Supplier Greiner Bio-One Supplier No. 653190
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for all applications for serological determination of HLA antigens. Greiner Bio-One TC Treated Terasaki Plates are supplied with fitting lid.

  • Manufactured from crystal clear polystyrene
  • TC surface treatment for improved cell adhesion
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 10 L (Well)
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Type Microtest Plate
Well Count 60
Includes Lid
Length (Metric) 83.3 mm
Width (Metric) 58 mm
Height (Metric) 11 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.