Learn More
Bio Rad Laboratories TC10 SYS SMPL SLIDE DUAL 30/PK
Catalog No. NC0167716
Encompass
Dual-chamber counting slide provides counts for two separate samples/ dilutions (10 ul each) and ensures accurate and reproducible counting. Includes 150 slides for up to 300 samples (145-0015)
Catalog No. NC0167716 Supplier Bio Rad Laboratories Supplier No. 1450011
Dual-chamber counting slide provides counts for two separate samples/ dilutions (10 ul each) and ensures accurate and reproducible counting. Includes 150 slides for up to 300 samples (145-0015)