Learn More
Huber USA TC100E-F, Immersion Cooler
Catalog No. 501108734
Encompass_Preferred
‐ Temperature range: -100°C ... +40°C - Cooling capacity (0°C): 0,16 kW‐ Cooling capacity (-80°C): 0,12 kW‐ Power supply: 115V 1~ 60Hz
Catalog No. 50-110-8734 Supplier Huber USA Supplier No. 3005.0065.99
Add to Cart
Add to Cart
Request a Quote
‐ Temperature range: -100°C ... +40°C - Cooling capacity (0°C): 0,16 kW‐ Cooling capacity (-80°C): 0,12 kW‐ Power supply: 115V 1~ 60Hz