Learn More
Medchemexpress LLC TC11 | 100823-03-8 | 99.6% | 10 MG
SDP
Catalog No. 5000338469
Encompass_Preferred
TC11 is an MCL1 degrader and a Caspase-9 and CDK1 activator. It functions as a phenylacetylamide derivative and is structurally related to immunomodulatory active molecules. TC11 induces the degradation of MCL1, leading to apoptotic death during prolonged mitotic arrest.
Catalog No. 50-003-38469 Supplier Medchemexpress LLC Supplier No. HY12947810MG
TC11 is an MCL1 degrader and a Caspase-9 and CDK1 activator. It functions as a phenylacetylamide derivative and is structurally related to immunomodulatory active molecules. TC11 induces the degradation of MCL1, leading to apoptotic death during prolonged mitotic arrest.