Learn More
Medchemexpress LLC HY-101524 100mg Medchemexpress, TC13172 CAS:2093393-05-4 Purity:>98%
SDP
Catalog No. 501932157
Encompass_Preferred
Medchemexpress, HY-101524 100mg TC13172 CAS:2093393-05-4 TC13172 is a mixed lineage kinase domain-like protein ( MLKL ) inhibitor with an EC 50 value of 2 nM for HT-29 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2157 Supplier Medchemexpress LLC Supplier No. HY101524100MG
Add to Cart
Add to Cart
Medchemexpress, HY-101524 100mg TC13172 CAS:2093393-05-4 TC13172 is a mixed lineage kinase domain-like protein ( MLKL ) inhibitor with an EC 50 value of 2 nM for HT-29 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.