Learn More
Bio Rad Laboratories TC20 Automated Cell Counter, 120-240 V
Catalog No. NC1119426
Encompass
120240 V, includes instrument, power supply, USB flash drive, 30 dual-chamber counting slides (60 counts), 1.5 mL trypan blue.
Catalog No. NC1119426 Supplier Bio Rad Laboratories Supplier No. 1450102
Add to Cart
Add to Cart
Request a Quote
120240 V, includes instrument, power supply, USB flash drive, 30 dual-chamber counting slides (60 counts), 1.5 mL trypan blue.