Learn More
Braintree Scientific TC9639 Thermal Cycler with multiformat block
SDP
Catalog No. 501956017
Encompass
Thermal Cycler, Intuitive touch screen control, Program Wizard - quick, easy & foolproof Unique multi-format block, 0.2ml tubes or strips (96), 0.5ml tubes (39), 96 well plate (1) Provides consistent, reliable results for laboratories with medium to high throughput. The program wizard and 10 preinstalled programs make it easier than ever to get up and running. A large, full color touch screen serves as the operations center where all functions are accessed. From the Home screen, simply touch an icon to view last program run. Enter the program library, or use the program wizard to quickly enter a protocol. In addition, icons for favorite programs can be added for faster access **Sold for research purposes only and not for human use**
Catalog No. 50-195-6017 Supplier Braintree Scientific Supplier No. T500096
 Restricted Item
Add to Cart
Add to Cart
Request a Quote
Thermal Cycler, Intuitive touch screen control, Program Wizard - quick, easy & foolproof Unique multi-format block, 0.2ml tubes or strips (96), 0.5ml tubes (39), 96 well plate (1) Provides consistent, reliable results for laboratories with medium to high throughput. The program wizard and 10 preinstalled programs make it easier than ever to get up and running. A large, full color touch screen serves as the operations center where all functions are accessed. From the Home screen, simply touch an icon to view last program run. Enter the program library, or use the program wizard to quickly enter a protocol. In addition, icons for favorite programs can be added for faster access **Sold for research purposes only and not for human use**