Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cayman Chemical TCA Assay Reagent (10%), 10 mL
SDP

Catalog No. 502895044
Encompass_Preferred

TCA Assay Reagent (10%), 10 mL

Catalog No. 50-289-5044 Supplier Cayman Chemical Supplier No. 70001610 ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCA Assay Reagent (10%), 10 mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.