Learn More
Cambridge Isotope Laboratories TCA CYCLE STND MIX 1
Supplier:  Cambridge Isotope Laboratories MSKTCA11
Encompass
TCA cycle standard mix 1 /cat no. MSK-TCA1/ is a single vial mix comprising 8 13C-labeled metabolites. These metabolites can function as labeled standards in the qualitative and/or quantitative exercises of MS metabolomics.
Catalog No. NC2943481