Learn More
Aviva Systems Biology Corp TCAP Antibody - middle region (ARP48228_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48228_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCAP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCAP. Alternate names: CMD1N, LGMD2G, T-cap, TELE, telethoninApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59767