Learn More
ABclonal Technology TCAP Rabbit mAb
SDP
Catalog No. 5000007305
Encompass_Preferred
Sarcomere assembly is regulated by the muscle protein titin Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere It serves as a scaffold to which myofibrils and other muscle related proteins are attached This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase interactions thought to be critical to sarcomere assembly Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G
Catalog No. 50-000-07305 Supplier ABclonal Technology Supplier No. A2273120UL
Sarcomere assembly is regulated by the muscle protein titin Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere It serves as a scaffold to which myofibrils and other muscle related proteins are attached This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase interactions thought to be critical to sarcomere assembly Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G