Learn More
Alkali Scientific TCBS Agar 20mL Plate, 10/Pk
Supplier: Alkali Scientific TTCP00
TCBS Agar is a selective and differential culture medium used for isolating Vibrio species from food, water, and clinical samples. The formulation contains high salt concentration and alkaline pH to suppress non target organisms while promoting Vibrio growth. It also includes sucrose and pH indicators that allow differentiation based on fermentation ability. Sucrose fermenting species produce yellow colonies, while non fermenting species appear green. This clear visual distinction helps identify important pathogens such as Vibrio cholerae. The 20mL plate format provides a ready to use solution for routine microbiological testing and environmental monitoring applications.