Learn More
NEOGEN Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar, 500 g
SDP
Catalog No. 502015303
Encompass_Preferred
Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar is used for the selective isolation of Vibrio cholerae and other enteropathogenic vibrios according to ISO 21872-1:2017 and ISO 21872-2:2007. Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar is not intended for use in the diagnosis of disease or other conditions in humans. TCBS Agar is also called Vibrio Selective Agar, and prepared according to the formula of Kobayashi et al. This formula is a modification of the selective medium from Nakanishi. All pathogenic Vibrio spp., except Vibrio hollisae, will grow on TCBS Agar. This highly selective agar meets the nutritional requirements of Vibrio spp. Vibrio spp. are able to grow in media containing increased salt concentrations, and some species are halophilic.For more information visit https://www.neogen.com/solutions/Microbiology/thiosulfate-citrate-bile-salts-sucrose-agar
Catalog No. 50-201-5303 Supplier NEOGEN Supplier No. 700003105
Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar is used for the selective isolation of Vibrio cholerae and other enteropathogenic vibrios according to ISO 21872-1:2017 and ISO 21872-2:2007. Thiosulfate Citrate Bile Salts Sucrose (TCBS) Agar is not intended for use in the diagnosis of disease or other conditions in humans. TCBS Agar is also called Vibrio Selective Agar, and prepared according to the formula of Kobayashi et al. This formula is a modification of the selective medium from Nakanishi. All pathogenic Vibrio spp., except Vibrio hollisae, will grow on TCBS Agar. This highly selective agar meets the nutritional requirements of Vibrio spp. Vibrio spp. are able to grow in media containing increased salt concentrations, and some species are halophilic.For more information visit https://www.neogen.com/solutions/Microbiology/thiosulfate-citrate-bile-salts-sucrose-agar