Learn More
Perkin Elmer US LLC TCD Add-On Kit - 120 V Clarus, Manual Pneumatics
Catalog No. 502113750
Encompass_Preferred
Kit includes all necessary items to install the TCD into the instrument:detector, heater, sensor, heater block, 1/16 in. gas line, and flow controller pneumatics. Requires, but does not include, amplifier (N6120015). The TCD can only be installed in the rear detector position.
Catalog No. 50-211-3750 Supplier Perkin Elmer US LLC Supplier No. N6520022
Add to Cart
Add to Cart
Request a Quote
Kit includes all necessary items to install the TCD into the instrument:detector, heater, sensor, heater block, 1/16 in. gas line, and flow controller pneumatics. Requires, but does not include, amplifier (N6120015). The TCD can only be installed in the rear detector position.