Learn More
Perkin Elmer US LLC TCD PPC Makeup Kit
Catalog No. 502113732
Encompass_Preferred
Includes parts required to add PPC controlled makeup gas to an existing TCD. Includes tubing, tee-piece, PPC pneumatics module, and PPC frit #4 (N6120155). Requires PPC capability in the GC.
Catalog No. 50-211-3732 Supplier Perkin Elmer US LLC Supplier No. N6120150
Includes parts required to add PPC controlled makeup gas to an existing TCD. Includes tubing, tee-piece, PPC pneumatics module, and PPC frit #4 (N6120155). Requires PPC capability in the GC.