Description
- Supports wet and dry PVC tubing in blood banking and laboratory workflows
- Uses single loading wafer technology for reliable sterile connection performance
- Helps maintain system integrity during component transfer and processing steps
- Designed for blood banks, cord blood facilities, cell therapy labs, and biopharma operations
- Supports process control where sterile connections and product protection are critical
Specifications
Specifications
| Product Type | Sterile Tube Welder |
| For Use With (Application) | Sterile Welding of PVC Tubing |