Description
- Copper wafer format supports reliable sterile tube welding performance
- Designed for use during PVC tubing connection procedures
- Supplied 200 wafers per case for routine operational inventory
- Supports sterile connection workflows in blood banking and cell therapy applications
- Essential consumable for facilities operating compatible TCD sterile tube welders
Specifications
Specifications
| Material | Copper |
| Product Type | Welding Wafer |
| For Use With (Application) | Sterile Tube Welding |
| For Use With (Equipment) | TCD Sterile Tube Welder |
| Includes | TCD Welding Wafers |
| Quantity | 200/Cs. |