Learn More
Perkin Elmer US LLC TCD with PPC Add-On Kit, 230 V
Catalog No. 502113718
Encompass_Preferred
Requires, but does not include, amplifier (N6120015). The TCD can only be installed in the rear detector position. The GC must be PPC ready. If not, a PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.
Catalog No. 50-211-3718 Supplier Perkin Elmer US LLC Supplier No. N6120125
Add to Cart
Add to Cart
Request a Quote
Requires, but does not include, amplifier (N6120015). The TCD can only be installed in the rear detector position. The GC must be PPC ready. If not, a PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.