Learn More
Perkin Elmer US LLC TCD/Flame Ionization Detector (FID) Series Operation Kit for PerkinElmer GCs
Catalog No. 502113654
Encompass_Preferred
TCD/FID Series Operation Kit used to direct sample effluent from the TCD to the FID.
Catalog No. 50-211-3654 Supplier Perkin Elmer US LLC Supplier No. N6120006
TCD/FID Series Operation Kit used to direct sample effluent from the TCD to the FID.