Learn More
Aviva Systems Biology Corp TCEA1 Antibody - N-terminal region (ARP38858_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38858_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCEA1. Alternate names: SII, TCEA, TF2S, GTF2S, TFIISApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53117