Learn More
Aviva Systems Biology Corp Tcea1 Antibody - N-terminal region (ARP37151_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37151_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tcea1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MGC103154, S-IIApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-589