Learn More
Aviva Systems Biology Corp TCEA1 Antibody - N-terminal region (ARP33407_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33407_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCEA1. Alternate names: SII, TCEA, TF2S, GTF2S, TFIISApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52905