Learn More
Aviva Systems Biology Corp Tcea2 Antibody - middle region (ARP36987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tcea2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Tcea2. Alternate names: AI326274, S-II-T1, SII-T1, TceatApplicationWB. Species reactivity: Horse, Mouse, Rat
Catalog No. 50-284-559