Learn More
Aviva Systems Biology Corp TCEA2 Antibody - N-terminal region (ARP58182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TCEA2. Alternate names: TFIISApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human
Catalog No. 50-286-451