Learn More
Aviva Systems Biology Corp TCEA3 Antibody - N-terminal region (ARP33409_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33409_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCEA3. Alternate names: TFIIS, TFIIS.HApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55005