Learn More
Aviva Systems Biology Corp TCEAL4 Antibody - middle region (ARP50341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEAL4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCEAL4. Alternate names: DKFZp686M0279, FLJ21174, NPD017ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60321