Learn More
Aviva Systems Biology Corp TCEAL6 Antibody - C-terminal region (ARP50342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEAL6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TCEAL6. Alternate names: Tceal3ApplicationWB. Species reactivity: Human
Catalog No. 50-285-505