Learn More
Aviva Systems Biology Corp Tceal7 Antibody - middle region (ARP50797_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50797_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tceal7. It was validated on Western Blot. . Alternate names: 1110018P05Rik, AI118095ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-285-541