Learn More
Aviva Systems Biology Corp ELOC Antibody - middle region (P100962_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100962_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCEB1. Alternate names: SIII, eloCApplicationWB. Species reactivity: Dog, Human, Zebrafish
Catalog No. 50-014-62804