Learn More
Aviva Systems Biology Corp ELOB Antibody - middle region (ARP31323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCEB2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TCEB2. Alternate names: SIII, ELOBApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54195