Learn More
ABclonal Technology TCEB2 Rabbit mAb
SDP
Catalog No. 5000007813
Encompass_Preferred
This gene encodes the protein elongin B which is a subunit of the transcription factor B (SIII) complex The SIII complex is composed of elongins A A2 B and C It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units Elongin A functions as the transcriptionally active component of the SIII complex whereas elongins B and C are regulatory subunits Elongin A2 is specifically expressed in the testis and capable of forming a stable complex with elongins B and C The von Hippel-Lindau tumor suppressor protein binds to elongins B and C and thereby inhibits transcription elongation Two alternatively spliced transcript variants encoding different isoforms have been described for this gene Pseudogenes have been identified on chromosomes 11 and 13
Catalog No. 50-000-07813 Supplier ABclonal Technology Supplier No. A21126100UL
This gene encodes the protein elongin B which is a subunit of the transcription factor B (SIII) complex The SIII complex is composed of elongins A A2 B and C It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units Elongin A functions as the transcriptionally active component of the SIII complex whereas elongins B and C are regulatory subunits Elongin A2 is specifically expressed in the testis and capable of forming a stable complex with elongins B and C The von Hippel-Lindau tumor suppressor protein binds to elongins B and C and thereby inhibits transcription elongation Two alternatively spliced transcript variants encoding different isoforms have been described for this gene Pseudogenes have been identified on chromosomes 11 and 13