Learn More
Chondrex Inc TCELL CHICK COL2, PDR, 0.5 MG
Catalog No. 501526675
Encompass_Preferred
0.5 mg (lyophilized) of 99% pure (by SDS-PAGE analysis) heat denatured chick type I collagen purfied from chick sternal cartilage. Intended for use as a stimulation antigen for T-cell proliferation assays and establishing type I collagen specific T-cell lines. Contact Chondrex, Inc. at support@chondrex.com for technical support.
Catalog No. 50-152-6675 Supplier Chondrex Inc Supplier No. 2021
0.5 mg (lyophilized) of 99% pure (by SDS-PAGE analysis) heat denatured chick type I collagen purfied from chick sternal cartilage. Intended for use as a stimulation antigen for T-cell proliferation assays and establishing type I collagen specific T-cell lines. Contact Chondrex, Inc. at support@chondrex.com for technical support.