Learn More
AdipoGen TCEP (5 g)
SDP
Catalog No. 502054661
Encompass_Preferred
Chemical. CAS: 51805-45-9. Formula: C9H15O6P·HCl. MW: 286.60. TCEP is an odorless, selective, and water-soluble strong reducing agent that is commonly used in biochemistry and molecular biology applications. It is commonly used to rapidly reduce protein and peptide disulfide bonds a preparatory step for gel electrophoresis. TCEP can be combined with proteases to simultaneously reduce and digest proteins prior to mass spectrometry in order to dramatically increase sequence coverage. More stable than DTT and useful in mass spectrometry applications. It has also been used to measure ascorbic acid and dehydroascorbic acid in biological samples. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).
Catalog No. 50-205-4661 Supplier AdipoGen Supplier No. CDXT0176G005
Chemical. CAS: 51805-45-9. Formula: C9H15O6P·HCl. MW: 286.60. TCEP is an odorless, selective, and water-soluble strong reducing agent that is commonly used in biochemistry and molecular biology applications. It is commonly used to rapidly reduce protein and peptide disulfide bonds a preparatory step for gel electrophoresis. TCEP can be combined with proteases to simultaneously reduce and digest proteins prior to mass spectrometry in order to dramatically increase sequence coverage. More stable than DTT and useful in mass spectrometry applications. It has also been used to measure ascorbic acid and dehydroascorbic acid in biological samples. TCEP is a useful chelating agent for various heavy metal ions as Zn(II), Cd(II), Pb(II), and Ni(II).