Learn More
Carbosynth LLC. Tris-(2-carboxyethyl)phosphine hydrochloride, 51805-45-9, MFCD00145469, 5 g
SDP
Supplier:  Carbosynth LLC. C18185G
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Formula: C9H16ClO6P, Molecular weight: 286.65 g/mol, 98%, Store at 5C, Synonyms: Tris-(2-carboxyethyl)phosphine HCl, Abbreviations: TCEP.
Catalog No. 50-121-6649