Learn More
Gold Biotechnology Inc TCEP-HCl 10 g
SDP
Catalog No. 501532843
Encompass_Preferred
TCEP (Tris (2-Carboxyethyl) phosphine Hydrochloride) is an odorless reducing agent suitable for protein applications and in the preparation for electrophoresis. TCEP is water soluble, is stable in a variety of solutions and is particularly useful in the labeling of cysteine residues with maleimides, preventing the cysteines from forming disulfide bonds.
Catalog No. 50-153-2843 Supplier Gold Biotechnology Inc Supplier No. TCEP10
TCEP (Tris (2-Carboxyethyl) phosphine Hydrochloride) is an odorless reducing agent suitable for protein applications and in the preparation for electrophoresis. TCEP is water soluble, is stable in a variety of solutions and is particularly useful in the labeling of cysteine residues with maleimides, preventing the cysteines from forming disulfide bonds.