Learn More
Selleck Chemical LLC TCEP Hydrochloride-100mg
SDP
Supplier:  Selleck Chemical LLC S4611100MG
Encompass_Preferred
TCEP (Tris(2-carboxyethyl)phosphine) hydrochloride, a non-thiol reducing agent, promotes NF-κB-DNA binding in a dose-related manner.Solutions are best fresh-prepared.
Catalog No. 50-238-9784