Learn More
Apexbio Technology LLC TCEP hydrochloride, 10g. Cas: 51805-45-9 MFCD: MFCD00145469. Reducing Agent
SDP
Supplier:  Apexbio Technology LLC B605510000
Encompass_Preferred
Tris(2-carboxyethyl)phosphine hydrochloride (TCEP HCL) is a water soluble strong reducing agent that cleave disulfide bonds. It is a non-thiol and non-volatile solid. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5372