Learn More
Selleck Chemical LLC TCEP Hydrochloride S4611-1g
SDP
Catalog No. 503110841
Encompass_Preferred
TCEP (Tris(2-carboxyethyl)phosphine) hydrochloride a non-thiol reducing agent promotes NF- B-DNA binding in a dose-related manner Solutions are best fresh-prepared
Catalog No. 50-311-0841 Supplier Selleck Chemical LLC Supplier No. S46111G
TCEP (Tris(2-carboxyethyl)phosphine) hydrochloride a non-thiol reducing agent promotes NF- B-DNA binding in a dose-related manner Solutions are best fresh-prepared