Learn More
AG Scientific Inc TCEP Hydrochloride, 5 G
Catalog No. 502237086
Encompass_Preferred
TCEP hydrochloride is a non-thiol reducing agent used to reduce and break disulfide bonds. TCEP HCl is widely preferred to the similar reducing agent DTT because of its greater effectiveness and resistance to oxidation from air exposure.CAS Number: 51805-45-9Molecular Weight: 286.6 DaChemical Formula: C9H15O6P HClSolubility: Soluble in waterStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7086 Supplier AG Scientific Inc Supplier No. T11805GM
TCEP hydrochloride is a non-thiol reducing agent used to reduce and break disulfide bonds. TCEP HCl is widely preferred to the similar reducing agent DTT because of its greater effectiveness and resistance to oxidation from air exposure.CAS Number: 51805-45-9Molecular Weight: 286.6 DaChemical Formula: C9H15O6P HClSolubility: Soluble in waterStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.