Learn More
Medchemexpress LLC TCEP hydrochloride | 51805-45-9 | 99.97% | 286.65 | 1 ML
SDP
Catalog No. 5000454952
Encompass_Preferred
TCEP hydrochloride is a non-thiol reducing agent that is more stable and produces a faster S-S reductive reaction than other chemical reductants. It is a trialkylphosphine that selectively reduces protein disulfides without altering the properties or interacting with thiol-directed agents in the reaction mixture. TCEP hydrochloride is also commonly used in DNA/AuNP chemistry.
  • Non-thiol reducing agent
  • More stable than other chemical reductants
  • Produces a faster S-S reductive reaction
  • Selectively reduces protein disulfides without altering properties or interacting with thiol-directed agents
  • Commonly used reducing agent in DNA/AuNP chemistry
Catalog No. 50-004-54952 Supplier Medchemexpress LLC Supplier No. HYW01150010MM/1ML
TCEP hydrochloride is a non-thiol reducing agent that is more stable and produces a faster S-S reductive reaction than other chemical reductants. It is a trialkylphosphine that selectively reduces protein disulfides without altering the properties or interacting with thiol-directed agents in the reaction mixture. TCEP hydrochloride is also commonly used in DNA/AuNP chemistry.
  • Non-thiol reducing agent
  • More stable than other chemical reductants
  • Produces a faster S-S reductive reaction
  • Selectively reduces protein disulfides without altering properties or interacting with thiol-directed agents
  • Commonly used reducing agent in DNA/AuNP chemistry