Learn More
Abcam TCEP hydrochloride (tris(2-carbo x yethyl)phosphine hydrochloride), non-thiol reducing reagent, 10G
Catalog No. 502583334
Encompass_Preferred
MW 286.64 Da, Purity >97%. Non-thiol reducing reagent. Reduces dehydroascorbic acid at low pH more efficiently than DTT (ab141390). Reductive at pH 1.5 to pH 9.0. Reverses Mg²+ inibition of NFκB-DNA binding.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3334 Supplier Abcam Supplier No. AB14204010G
MW 286.64 Da, Purity >97%. Non-thiol reducing reagent. Reduces dehydroascorbic acid at low pH more efficiently than DTT (ab141390). Reductive at pH 1.5 to pH 9.0. Reverses Mg²+ inibition of NFκB-DNA binding.
The product is subject to the following: Abcam Restricted Use Statement