Learn More
Ubpbio TCEP SOLUTION (1M), 10ML
Supplier:  Ubpbio P102110
Encompass_Preferred
Protein Purification Reagents – TCEP solution (1M) (UBPBio catalog# P1021-10, P1020-100) – TCEP (tris(2-carboxyethyl)phosphine) is a strong reducing agent. Compared to dithiothreitol (DTT) and β-mercaptoethanol, TCEP has the advantages of being odorless, a more powerful reducing agent, an irreversible reducing agent, more hydrophilic, and more resistant to oxidation in air. It is soluble in water and stable at neutral pH. It does not reduce metals used in immobilized metal affinity chromatography such as Ni-NTA resin. TCEP is particularly useful when labeling cysteine residues with maleimides.
Catalog No. 50-112-6937