Learn More
Aviva Systems Biology Corp TCERG1 Antibody - middle region (ARP32830_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32830_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCERG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCERG1. Alternate names: CA150, MGC133200, TAF2S, Urn1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54780