Learn More
ABclonal Technology TCERG1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148501000UL
Encompass_Preferred
This gene encodes a nuclear protein that regulates transcriptional elongation and pre-mRNA splicing. The encoded protein interacts with the hyperphosphorylated C-terminal domain of RNA polymerase II via multiple FF domains, and with the pre-mRNA splicing factor SF1 via a WW domain. Alternative splicing results in multiple transcripts variants encoding different isoforms.
Catalog No. 50-001-05095