Learn More
Santa Cruz Biotechnology TCF-4 Antibody (D-4): sc-166699, 200 µg/ml
Catalog No. NC3313337
Encompass
TCF-4 Antibody (D-4) is a mouse monoclonal IgG1 κ TCF-4 antibody, cited in 49 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 565-592 at the C-terminus of TCF-4 of human originTCF-4 Antibody (D-4) is recommended for detection of TCF-4 of mouse, rat and human origin by WB, IP, IF and ELISAAnti-TCF-4 Antibody (D-4) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM
Catalog No. NC3313337 Supplier Santa Cruz Biotechnology Supplier No. SC166699
TCF-4 Antibody (D-4) is a mouse monoclonal IgG1 κ TCF-4 antibody, cited in 49 publications, provided at 200 µg/mlspecific for an epitope mapping between amino acids 565-592 at the C-terminus of TCF-4 of human originTCF-4 Antibody (D-4) is recommended for detection of TCF-4 of mouse, rat and human origin by WB, IP, IF and ELISAAnti-TCF-4 Antibody (D-4) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM