Learn More
Cell Signaling Technology TCF1/TCF7 Monoclonal Antibody, Rabbit, BSA and Azide Free, 100 μg
SDP
Catalog No. NC2185519
Encompass
This TCF1/TCF7 (C63D9) Rabbit Monoclonal Antibody is a carrier-free formulation designed to detect endogenous levels of total TCF1/TCF7 protein. Produced by immunizing animals with a synthetic peptide corresponding to a region surrounding Pro96 of the human TCF1/TCF7 protein, this antibody does not recognize dominant negative isoforms lacking the amino-terminal β-catenin binding domain and does not cross-react with LEF1. This antibody is ideal for specialized or custom labeling applications.
  • Validated for western blotting, immunohistochemistry (paraffin), immunofluorescence (frozen and immunocytochemistry), and flow cytometry (fixed/permeabilized)
  • Supplied in 1X PBS without BSA or sodium azide
  • Quality control tested by size exclusion chromatography
  • Reactivity with human and mouse samples
  • Recommended to store at -20°C in single-use aliquots
This item is not returnable. View return policy
Catalog No. NC2185519 Supplier Cell Signaling Technology Supplier No. 85942SF
This TCF1/TCF7 (C63D9) Rabbit Monoclonal Antibody is a carrier-free formulation designed to detect endogenous levels of total TCF1/TCF7 protein. Produced by immunizing animals with a synthetic peptide corresponding to a region surrounding Pro96 of the human TCF1/TCF7 protein, this antibody does not recognize dominant negative isoforms lacking the amino-terminal β-catenin binding domain and does not cross-react with LEF1. This antibody is ideal for specialized or custom labeling applications.
  • Validated for western blotting, immunohistochemistry (paraffin), immunofluorescence (frozen and immunocytochemistry), and flow cytometry (fixed/permeabilized)
  • Supplied in 1X PBS without BSA or sodium azide
  • Quality control tested by size exclusion chromatography
  • Reactivity with human and mouse samples
  • Recommended to store at -20°C in single-use aliquots