Learn More
Aviva Systems Biology Corp TCF12 Antibody - middle region (P100772_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100772_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCF12. Alternate names: HEB, HTF4, HsT17266, bHLHb20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62706